2-Furohydrazide 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Furoic acid hydrazide, 2-Furoylhydrazine, 2-Furancarboxylic acid Hydrazide, 2-Furoic hydrazide, 2-Furoic acid Hydrazide, 2-Furancarboxylic acid hydrazide, 2-Furoic Hydrazide

A13630.09 A13630.18 A13630.30
ALFAA13630.09EA 0 SGD
ALFAA13630.09 ALFAA13630.18 ALFAA13630.30
2-Furohydrazide 98
2-Furohydrazide

Purity: 98% CAS No.: 3326-71-4 Molecular Formula: C5H6N2O2 Formula Weight: 126.12 Melting Point: 77-81° Beilstein Reference: 114435

Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00003235
CAS Number: 3326-71-4
EINECS: 222-046-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR