DL-Tyrosine ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Hydroxyphenylalanine, 3-(p-Hidroxifenil)-DL-alanina, H-DL-Tyr-OH

A13740.09 A13740.18 A13740.30
ALFAA13740.09EA 0 SGD
ALFAA13740.09 ALFAA13740.18 ALFAA13740.30
DL-Tyrosine ≥98%
DL-Tyrosine

Purity: 98% CAS No.: 556-03-6 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Melting Point: ca 325° dec. Beilstein Reference: 515881 Merck Reference: 14,9839 MDL No.: MFCD00063074

Formula: C₉H₁₁NO₃
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: ∼ 325 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00063074
CAS Number: 556-03-6
EINECS: 209-113-1
Merck Index: 14,9839

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR