Ammonium heptamolybdate tetrahydrate ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Hexaammonium heptamolybdate tetrahydrate, Ammonium molybdate(VI) tetrahydrate, Ammonium molybdate tetrahydrate, Hexammonium heptamolybdate 4-hydrate, Molybdic acid ammonium salt tetrahydrate

A13766.0B A13766.18 A13766.30
ALFAA13766.0BEA 0 SGD
ALFAA13766.0B ALFAA13766.18 ALFAA13766.30
Ammonium heptamolybdate tetrahydrate ≥99%
Ammonium heptamolybdate tetrahydrate

Purity: 99% CAS No.: 12054-85-2 Molecular Formula: (NH4)6Mo7O24.4H2O Formula Weight: 1235.86 (1163.79anhy) Melting Point: ca 190° dec. Merck Reference: 14,533 MDL No.: MFCD00167059

Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Melting Pt: ∼ 190 °C (dec.)
Density: 2,498
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 14,533

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR