Caproic acid 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Hexanoic acid

A13789.0F A13789.AE A13789.AP
ALFAA13789.0FEA 0 SGD
ALFAA13789.0F ALFAA13789.AE ALFAA13789.AP
Caproic acid 98
Caproic acid

Purity: 98+% CAS No.: 142-62-1 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 104°(219°F) Physical Form: Clear liquid Density: 0.929 Refractive Index: 1.4165 Appearance: Pale yellow Melting Point: -4° Boiling Point: 202-203° Beilstein Reference: 773837 Merck Reference: 14,1759 UN No.: 2829 MDL No.: MFCD00004421

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 202…203 °C
Melting Pt: –4 °C
Density: 0.929
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004421
CAS Number: 142-62-1
EINECS: 205-550-7
UN: 2829
ADR: 8 III
Merck Index: 13,01765

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR