2-Bromopyrimidine ≥98%

Supplier: Thermo Fisher Scientific
Warning

A13791.14 A13791.03 A13791.06 A13791.22
ALFAA13791.14EA 0 SGD
ALFAA13791.14 ALFAA13791.03 ALFAA13791.06 ALFAA13791.22
2-Bromopyrimidine ≥98%
2-Bromopyrimidine

Purity: 98+% CAS No.: 4595-60-2 Molecular Formula: C4H3BrN2 Formula Weight: 158.99 Storage Temperature: 2°C to 8°C Melting Point: 53-57° Boiling Point: 62-64°/1.5mm Beilstein Reference: 605945 MDL No.: MFCD00014601

Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 62…64 °C (1.5 mmHg)
Melting Pt: 53…57 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00014601
CAS Number: 4595-60-2
EINECS: 224-993-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR