2-Chloro-6-methylnicotinic acid 97

Supplier: Thermo Fisher Scientific
Warning

A13857.06 A13857.14 A13857.22
ALFAA13857.06EA 0 SGD
ALFAA13857.06 ALFAA13857.14 ALFAA13857.22
2-Chloro-6-methylnicotinic acid 97
2-Chloro-6-methylnicotinic acid

Purity: 97% CAS No.: 30529-70-5 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: 157-162° Beilstein Reference: 473070

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 157…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00134165
CAS Number: 30529-70-5
EINECS: 250-229-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR