Ethyl-2-furoate 99

Supplier: Thermo Fisher Scientific

Synonyms: Ethyl2-furoate, Ethyl furan-2-carboxylate

A13909.14 A13909.22 A13909.36
ALFAA13909.14EA 0 SGD
ALFAA13909.14 ALFAA13909.22 ALFAA13909.36
Ethyl-2-furoate 99
Ethyl 2-furoate

Purity: 99% CAS No.: 614-99-3 Molecular Formula: C7H8O3 Formula Weight: 140.14 Storage Temperature: 2°C to 8°C Flash Point: 70°(158°F) Melting Point: 31-36° Boiling Point: 195-196° Beilstein Reference: 2653 Merck Reference: 14,4307 MDL No.: MFCD00003237

Formula: C₇H₈O₃
MW: 140.14 g/mol
Boiling Pt: 195…196 °C
Melting Pt: 31…36 °C
Density: 1.120
Flash Pt: 70 °C (158 °F)
MDL Number: MFCD00003237
CAS Number: 614-99-3
EINECS: 210-404-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR