3-Chlorobenzoylacetonitrile ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3'-Chloro-2-cyanoacetophenone, (3-Chlorobenzoyl)acetonitrile, 3-(3-Chlorophenyl)-3-oxopropanenitrile

A13913.03 A13913.06
ALFAA13913.03EA 0 SGD
ALFAA13913.03 ALFAA13913.06
3-Chlorobenzoylacetonitrile ≥98%
3-Chlorobenzoylacetonitrile

Purity: 98% CAS No.: 21667-62-9 Molecular Formula: C9H6ClNO Formula Weight: 179.61 Melting Point: 78-83° Beilstein Reference: 2414476

Formula: C₉H₆ClNO
MW: 179.61
Melting Pt: 78…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00067891
CAS Number: 21667-62-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR