4'-Bromo-3-chloropropiophenone ≥94%

Supplier: Thermo Fisher Scientific
Warning

A13988.06 A13988.09 A13988.18 A13988.30
ALFAA13988.06EA 0 SGD
ALFAA13988.06 ALFAA13988.09 ALFAA13988.18 ALFAA13988.30
4'-Bromo-3-chloropropiophenone ≥94%
4'-Bromo-3-chloropropiophenone

Purity: 94% CAS No.: 31736-73-9 Molecular Formula: C9H8BrClO Formula Weight: 247.53 Melting Point: 57-62° Beilstein Reference: 1944367

Formula: C₉H₈BrClO
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 57…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00018979
CAS Number: 31736-73-9
EINECS: 250-784-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR