1-Bromo-3-chlorobenzene ≥99%

Supplier: Thermo Fisher Scientific
Warning

A14026.22 A14026.36
ALFAA14026.22EA 129 SGD
ALFAA14026.22 ALFAA14026.36
1-Bromo-3-chlorobenzene ≥99%
1-Bromo-3-chlorobenzene

Purity: 99% CAS No.: 108-37-2 Molecular Formula: C6H4BrCl Formula Weight: 191.46 Flash Point: 80°(176°F) Refractive Index: 1.5770 Odour: Aromatic Melting Point: -22° Boiling Point: 195-196° Beilstein Reference: 1446353 MDL No.: MFCD00000568

Formula: C₆H₄BrCl
Boiling Pt: 195…196 °C
Melting Pt: –22 °C
Density: 1,630
Flash Pt: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000568
CAS Number: 108-37-2
EINECS: 203-575-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR