1-Ethyl-4-iodobenzene ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Ethyliodobenzene

A14138.09 A14138.18
ALFAA14138.09EA 0 SGD
ALFAA14138.09 ALFAA14138.18
1-Ethyl-4-iodobenzene ≥98%
1-Ethyl-4-iodobenzene

Purity: 98% CAS No.: 25309-64-2 Molecular Formula: C8H9I Formula Weight: 232.07 Flash Point: 107°(224°F) Refractive Index: 1.5910 Odour: Mild Melting Point: -17° Boiling Point: 112-113°/20mm Beilstein Reference: 2076973 MDL No.: MFCD00060663

Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 112…113 °C (20 mmHg)
Melting Pt: –17 °C
Density: 1.600
Flash Pt: 107 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060663
CAS Number: 25309-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR