2,5-Dichlorobenzonitrile ≥98%

Supplier: Thermo Fisher Scientific
Danger

A14141.06 A14141.14
ALFAA14141.06EA 0 SGD
ALFAA14141.06 ALFAA14141.14
2,5-Dichlorobenzonitrile ≥98%
2,5-Dichlorobenzonitrile

Purity: 98% CAS No.: 21663-61-6 Molecular Formula: C7H3Cl2N Formula Weight: 172.01 Melting Point: 128-131° Beilstein Reference: 2045293 UN No.: 3439 MDL No.: MFCD00019741

Formula: C₇H₃Cl₂N
MW: 172.01 g/mol
Boiling Pt: 269
Melting Pt: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00019741
CAS Number: 21663-61-6
EINECS: 244-518-7
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR