Ethyl phenoxyacetate ≥99%

Supplier: Thermo Fisher Scientific

A14156.09 A14156.18 A14156.30
ALFAA14156.09EA 0 SGD
ALFAA14156.09 ALFAA14156.18 ALFAA14156.30
Ethyl phenoxyacetate ≥99%
Ethyl phenoxyacetate

Purity: 99% CAS No.: 2555-49-9 Molecular Formula: C6H5OCH2CO2CH2CH3 Formula Weight: 180.20 Flash Point: 110°(230°F) Refractive Index: 1.5055 Boiling Point: 251-253° Beilstein Reference: 1871712 MDL No.: MFCD00026895

Formula: C₁₀H₁₂O₃
Boiling Pt: 251…253 °C
Density: 1,108
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026895
CAS Number: 2555-49-9
EINECS: 219-867-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR