Ethoxyacetylene 50% (w/w) in hexane

Supplier: Thermo Fisher Scientific
Danger

Synonyms: ethynoxyethane, Ethyl ethynyl ether, ethoxy-ethyne

A14245.03 A14245.06 A14245.14
ALFAA14245.03EA 0 SGD
ALFAA14245.03 ALFAA14245.06 ALFAA14245.14
Ethoxyacetylene 50% (w/w) in hexane
Ethoxyacetylene

CAS No.: 927-80-0 Molecular Formula: C4H6O Formula Weight: 70.09 Storage Temperature: 2°C to 8°C Flash Point: -28°(-18°F) Refractive Index: 1.3800 Odour: Recognizable Boiling Point: 55-56° Beilstein Reference: 741882 UN No.: UN1993 MDL No.: MFCD00009247

Formula: C₄H₆O
MW: 70.091 g/mol
Boiling Pt: 55…56 °C
Density: 0.718
Flash Pt: –28 °C (–18 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009247
CAS Number: 927-80-0
EINECS: 213-164-5
UN: 1993
ADR: 3 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR