4-(Hydroxymethyl)benzyl alcohol ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: p-Phenylenedimethanol, 1,4-Benzenedimethanol, 1,4-Bis(hydroxymethyl)-benzene, p-Xylene-α,α'-diol

A14405.06 A14405.14 A14405.22
ALFAA14405.06EA 0 SGD
ALFAA14405.06 ALFAA14405.14 ALFAA14405.22
4-(Hydroxymethyl)benzyl alcohol ≥99%
4-(Hydroxymethyl)benzyl alcohol

Purity: 99% CAS No.: 589-29-7 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 117-121° Boiling Point: 138-143°/1mm MDL No.: MFCD00004665

Formula: C₈H₁₀O₂
Boiling Pt: 138…143 °C (1 mmHg)
Melting Pt: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00004665
CAS Number: 589-29-7
EINECS: 209-641-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR