Ethyl-3-aminobenzoate 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ethyl 3-aminobenzoate

A14448.09 A14448.18 A14448.22 A14448.30
ALFAA14448.09EA 0 SGD
ALFAA14448.09 ALFAA14448.18 ALFAA14448.22 ALFAA14448.30
Ethyl-3-aminobenzoate 98%
Ethyl 3-aminobenzoate

Purity: 98% CAS No.: 582-33-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Flash Point: 110°(230°F) Refractive Index: 1.5610 Melting Point: 27-30° Boiling Point: 294° Beilstein Reference: 1101967 MDL No.: MFCD00007794

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 294 °C
Melting Pt: 27…30 °C
Density: 1.106
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007794
CAS Number: 582-33-2
EINECS: 209-482-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR