2-Fluoro-6-phenoxybenzonitrile 95%

Supplier: Thermo Fisher Scientific
Warning

A14566.06
ALFAA14566.06EA 0 SGD
ALFAA14566.06
2-Fluoro-6-phenoxybenzonitrile 95%
2-Fluoro-6-phenoxybenzonitrile

Purity: 95% CAS No.: 175204-06-5 Molecular Formula: C13H8FNO Formula Weight: 213.21 Melting Point: 90-93° MDL No.: MFCD00068201

Formula: C₁₃H₈FNO
Boiling Pt: 338 °C (1013 hPa)
Melting Pt: 90…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00068201
CAS Number: 175204-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR