N-Phenylmaleimide ≥98%

Supplier: Thermo Fisher Scientific
Danger

A14616.14 A14616.22 A14616.36
ALFAA14616.14EA 0 SGD
ALFAA14616.14 ALFAA14616.22 ALFAA14616.36
N-Phenylmaleimide ≥98%
N-Phenylmaleimide

Purity: 98+% CAS No.: 941-69-5 Molecular Formula: C10H7NO2 Formula Weight: 173.17 Melting Point: 87-91° Boiling Point: 162-163°/12mm Merck Reference: 14,7299 UN No.: 2811 MDL No.: MFCD00005502

Formula: C₁₀H₇NO₂
MW: 173.17
Boiling Pt: 162…163 °C (12 mmHg)
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00005502
CAS Number: 941-69-5
EINECS: 213-382-0
UN: 2811
ADR: 6.1 III
Merck Index: 12,07452

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR