4-Bromo-o-xylene ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Bromo-1,2-dimethylbenzene

A14736.06 A14736.14 A14736.22
ALFAA14736.06EA 0 SGD
ALFAA14736.06 ALFAA14736.14 ALFAA14736.22
4-Bromo-o-xylene ≥97%
4-Bromo-o-xylene

Purity: 97% CAS No.: 583-71-1 Molecular Formula: C8H9Br Formula Weight: 185.07 Flash Point: 78°(172°F) Refractive Index: 1.5560 Boiling Point: 214-215° Beilstein Reference: 2040354 MDL No.: MFCD00000086

Formula: C₈H₉Br
Boiling Pt: 214…215 °C
Density: 1,370
Flash Pt: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000071
CAS Number: 583-71-1
EINECS: 209-518-3
UN: 1701
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR