2-Chlorobenzoyl chloride ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: o-Chlorobenzoyl chloride

A14785.0B A14785.22 A14785.30
ALFAA14785.0BEA 0 SGD
ALFAA14785.0B ALFAA14785.22 ALFAA14785.30
2-Chlorobenzoyl chloride ≥97%
2-Chlorobenzoyl chloride

Purity: 97% CAS No.: 609-65-4 Molecular Formula: C7H4Cl2O Formula Weight: 175.01 Flash Point: 125°(257°F) Refractive Index: 1.5718 Odour: Acrid Melting Point: -4° Boiling Point: 237-239° Beilstein Reference: 386435 UN No.: 3265 MDL No.: MFCD00000660

Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 237…239 °C
Melting Pt: –4 °C
Density: 1.379
Flash Pt: 125 °C (257 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000660
CAS Number: 609-65-4
EINECS: 210-194-0
UN: 3265
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR