3-Bromo-4-fluorobenzaldehyde ≥98%

Supplier: Thermo Fisher Scientific
Warning

A15198.06 A15198.14
ALFAA15198.06EA 0 SGD
ALFAA15198.06 ALFAA15198.14
3-Bromo-4-fluorobenzaldehyde ≥98%
3-Bromo-4-fluorobenzaldehyde

Purity: 98% CAS No.: 77771-02-9 Molecular Formula: C7H4BrFO Formula Weight: 203.01 Flash Point: 110°(230°F) Refractive Index: 1.5740 Melting Point: 29-33° Boiling Point: 138-139°/2.5mm Beilstein Reference: 5806226 MDL No.: MFCD00042186

Formula: C₇H₄BrFO
Boiling Pt: 138…139 °C (2,5 mmHg)
Melting Pt: 29…33 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042186
CAS Number: 77771-02-9
EINECS: 278-764-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR