α-Bromo-m-toluonitrile ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-(Bromomethyl)benzonitrile, m-(Bromomethyl)benzonitrile

A15244.06 A15244.14
ALFAA15244.06EA 0 SGD
ALFAA15244.06 ALFAA15244.14
α-Bromo-m-toluonitrile ≥98%
α-Bromo-m-toluonitrile

Purity: 98% CAS No.: 28188-41-2 Molecular Formula: C8H6BrN Formula Weight: 196.05 Melting Point: 92-96° Beilstein Reference: 742356 UN No.: 2923 MDL No.: MFCD00001809

Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 130 °C (4 mmHg)
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00001809
CAS Number: 28188-41-2
EINECS: 248-890-1
UN: 2923
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR