3-Nitrophenol ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: m-Nitrophenol

A15312.09
ALFAA15312.09EA 0 SGD
ALFAA15312.09
3-Nitrophenol ≥98%
3-Nitrophenol

Purity: 98+% CAS No.: 554-84-7 Molecular Formula: C6H5NO3 Formula Weight: 139.11 Flash Point: 100°(212°F) Melting Point: 93-98° Boiling Point: 194°/70mm Beilstein Reference: 1907946 Merck Reference: 14,6619 UN No.: 1663 MDL No.: MFCD00007240

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 194 °C (70 mmHg)
Melting Pt: 93…98 °C
Density: 1.490
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007240
CAS Number: 554-84-7
EINECS: 209-073-5
UN: 1663
ADR: 6.1 III
Merck Index: 13,06652

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR