3-Tolualdehyde ≥97% stabilised

Supplier: Thermo Fisher Scientific
Warning

Synonyms: m-Tolualdehyde, 3-Methylbenzaldehyde, m-Tolylaldehyde

Stabilization: Stabilised with hydroquinone (HQ)

A15313.14 A15313.06 A15313.22
ALFAA15313.14EA 0 SGD
ALFAA15313.14 ALFAA15313.06 ALFAA15313.22
3-Tolualdehyde ≥97% stabilised
3-Tolualdehyde

Stabilized with 0.1% hydroquinone


Purity: 97% CAS No.: 620-23-5 Molecular Formula: C8H8O Formula Weight: 120.15 Flash Point: 78°(172°F) Refractive Index: 1.5410 Odour: Almond-like Boiling Point: 198-200° Beilstein Reference: 741964 MDL No.: MFCD00003374

Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 198…200 °C
Density: 1.020
Flash Pt: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003374
CAS Number: 620-23-5
EINECS: 210-632-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR