4-Bromo-m-toluidine ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-Amino-6-bromotoluene, 4-Bromo-3-methylaniline, 5-Amino-2-bromotoluene

A15417.09 A15417.18
ALFAA15417.09EA 0 SGD
ALFAA15417.09 ALFAA15417.18
4-Bromo-m-toluidine ≥97%
4-Bromo-m-toluidine

Purity: 97% CAS No.: 6933-10-4 Molecular Formula: C7H8BrN Formula Weight: 186.06 Odour: Characteristic Melting Point: 79-83° Boiling Point: 239-241° Beilstein Reference: 2689600 UN No.: 2811 MDL No.: MFCD00007828

Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 239…241 °C
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00007828
CAS Number: 6933-10-4
EINECS: 230-056-3
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR