2-Bromo-5-nitrotoluene ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Bromo-2-methyl-4-nitrobenzene

A15450.18
ALFAA15450.18EA 0 SGD
ALFAA15450.18
2-Bromo-5-nitrotoluene ≥98%
2-Bromo-5-nitrotoluene

Purity: 98% CAS No.: 7149-70-4 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Melting Point: 76-79° Beilstein Reference: 1940342

Formula: C₇H₆BrNO₂
Boiling Pt: 140…143 °C (17 mmHg)
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00007281
CAS Number: 7149-70-4
EINECS: 230-481-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR