2-Ethoxybenzyl alcohol 98

Supplier: Thermo Fisher Scientific

Synonyms: (2-Ethoxyphenyl)methanol

A15477.14 A15477.22
ALFAA15477.14EA 0 SGD
ALFAA15477.14 ALFAA15477.22
2-Ethoxybenzyl alcohol 98
2-Ethoxybenzyl alcohol

Purity: 98% CAS No.: 71672-75-8 Molecular Formula: C9H12O2 Formula Weight: 152.19 Flash Point: 110°(230°F) Refractive Index: 1.5320 Boiling Point: 263-265° Beilstein Reference: 2555374 MDL No.: MFCD00004616

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 263…265 °C
Melting Pt: 0
Density: 1.074
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004616
CAS Number: 71672-75-8
EINECS: 275-811-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR