2-Nitrobenzyl alcohol 97

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (2-Nitrophenyl)methanol

A15496.14 A15496.22 A15496.36
ALFAA15496.14EA 0 SGD
ALFAA15496.14 ALFAA15496.22 ALFAA15496.36
2-Nitrobenzyl alcohol 97
2-Nitrobenzyl alcohol

Purity: 97% CAS No.: 612-25-9 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: 70-74° Boiling Point: 270° MDL No.: MFCD00007186

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 270 °C
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00007186
CAS Number: 612-25-9
EINECS: 210-302-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR