4-Methoxybenzyl alcohol ≥98%

Supplier: Thermo Fisher Scientific
Danger

A15559.0E A15559.22 A15559.36
ALFAA15559.0EEA 0 SGD
ALFAA15559.0E ALFAA15559.22 ALFAA15559.36
4-Methoxybenzyl alcohol ≥98%
4-Methoxybenzyl alcohol

Purity: 98% CAS No.: 105-13-5 Molecular Formula: C8H10O2 Formula Weight: 138.17 Storage Temperature: 2°C to 8°C Flash Point: 113°(235°F) Refractive Index: 1.5450 Melting Point: 22-25° Boiling Point: 259° Beilstein Reference: 636654 Merck Reference: 14,604 MDL No.: MFCD00004653

Formula: C₈H₁₀O₂
Boiling Pt: 259 °C
Melting Pt: 22…25 °C
Density: 1,110
Flash Pt: 113 °C (235 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004653
CAS Number: 105-13-5
EINECS: 203-273-6
Merck Index: 14,604

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR