Lanthanum(III) chloride heptahydrate ≥99%

Supplier: Thermo Fisher Scientific
Warning

A15575.0E A15575.22 A15575.36
ALFAA15575.0EEA 0 SGD
ALFAA15575.0E ALFAA15575.22 ALFAA15575.36
Lanthanum(III) chloride heptahydrate ≥99%
Lanthanum (III) chloride heptahydrate

Purity: 99% CAS No.: 10025-84-0 Molecular Formula: LaCl3.7H2O Formula Weight: 371.37 (245.27anhy) Odour: Odourless Melting Point: 91° dec. MDL No.: MFCD00149756

Formula: LaCl₃·7H₂O
Boiling Pt: ∼860 °C (1013 hPa)
Melting Pt: 91 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0
EINECS: 233-237-5
Merck Index: 14,5363

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR