N-(3-Bromopropyl)phthalimide ≥98%

Supplier: Thermo Fisher Scientific

A15587.14 A15587.22 A15587.36
ALFAA15587.14EA 0 SGD
ALFAA15587.14 ALFAA15587.22 ALFAA15587.36
N-(3-Bromopropyl)phthalimide ≥98%
N-(3-Bromopropyl)phthalimide

Purity: 98% CAS No.: 5460-29-7 Molecular Formula: C11H10BrNO2 Formula Weight: 268.12 Odour: Odourless Melting Point: 72-75° Beilstein Reference: 157547 MDL No.: MFCD00005904

Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
Boiling Pt: 433
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00005904
CAS Number: 5460-29-7
EINECS: 226-738-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR