2-Bromo-1,4-dimethoxybenzene ≥98%

Supplier: Thermo Fisher Scientific
Warning

A15640.09 A15640.18 A15640.30
ALFAA15640.09EA 0 SGD
ALFAA15640.09 ALFAA15640.18 ALFAA15640.30
2-Bromo-1,4-dimethoxybenzene ≥98%
2-Bromo-1,4-dimethoxybenzene

Purity: 98% CAS No.: 25245-34-5 Molecular Formula: C8H9BrO2 Formula Weight: 217.07 Flash Point: 110°(230°F) Refractive Index: 1.5700 Boiling Point: 260-262° Beilstein Reference: 2441884 MDL No.: MFCD00008355

Formula: C₈H₉BrO₂
Boiling Pt: 260…262 °C
Density: 1,441
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008355
CAS Number: 25245-34-5
EINECS: 246-756-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR