Methyl anthranilate ≥99%

Supplier: Thermo Fisher Scientific
Warning

A15681.0B A15681.30 A15681.36
ALFAA15681.0BEA 0 SGD
ALFAA15681.0B ALFAA15681.30 ALFAA15681.36
Methyl anthranilate ≥99%
Methyl anthranilate

Purity: 99% CAS No.: 134-20-3 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Flash Point: 123°(253°F) Physical Form: Liquid Density: 1.170 Refractive Index: 1.5830 Appearance: Colourless to pale yellow Melting Point: 24° Boiling Point: 260° Beilstein Reference: 606965 Merck Reference: 14,6020 MDL No.: MFCD00007710

Formula: C₈H₉NO₂
Boiling Pt: 260 °C
Melting Pt: 24 °C
Density: 1,170
Flash Pt: 123 °C (253 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007710
CAS Number: 134-20-3
EINECS: 205-132-4
Merck Index: 14,6020

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR