6-Bromocaproic acid 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 6-Bromohexanoic acid

A15761.14 A15761.22 A15761.36
ALFAA15761.14EA 0 SGD
ALFAA15761.14 ALFAA15761.22 ALFAA15761.36
6-Bromocaproic acid 98
6-Bromocaproic acid

Purity: 98+% CAS No.: 4224-70-8 Molecular Formula: Br(CH2)5CO2H Formula Weight: 195.06 Flash Point: 95°(203°F) Odour: Odourless Boiling Point: 165-170°/20mm Beilstein Reference: 1749740 UN No.: 3261 MDL No.: MFCD00004422

Formula: BrH₂C(CH₂)₄COOH
MW: 195.06 g/mol
Boiling Pt: 165…170 °C (20 mmHg)
Melting Pt: 32…35 °C
Density: 1.4068 g/cm³ (30 °C)
Flash Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004422
CAS Number: 4224-70-8
EINECS: 224-176-5
UN: 3261

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR