3-Nitrobenzonitrile ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: m-Nitrobenzonitrile

A15821.09 A15821.14 A15821.30
ALFAA15821.09EA 0 SGD
ALFAA15821.09 ALFAA15821.14 ALFAA15821.30
3-Nitrobenzonitrile ≥98%
3-Nitrobenzonitrile

Purity: 98% CAS No.: 619-24-9 Molecular Formula: C7H4N2O2 Formula Weight: 148.12 Odour: Almond-like Melting Point: 114-117° Boiling Point: 165°/21mm Beilstein Reference: 637674 UN No.: 3439 MDL No.: MFCD00007194

Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Boiling Pt: 165 °C (21 mmHg)
Melting Pt: 114…117 °C
Density: 0.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007194
CAS Number: 619-24-9
EINECS: 210-587-7
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR