3-Bromo-o-xylene ≥99%

Supplier: Thermo Fisher Scientific
Warning

A15889.06 A15889.14 A15889.22
ALFAA15889.06EA 0 SGD
ALFAA15889.06 ALFAA15889.14 ALFAA15889.22
3-Bromo-o-xylene ≥99%
3-Bromo-o-xylene

Purity: 99% CAS No.: 576-23-8 Molecular Formula: C8H9Br Formula Weight: 185.07 Flash Point: 80°(176°F) Refractive Index: 1.5600 Boiling Point: 212-214° Beilstein Reference: 2077312 MDL No.: MFCD00000069

Formula: C₈H₉Br
Boiling Pt: 212…214 °C
Density: 1,365
Flash Pt: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000069
CAS Number: 576-23-8
EINECS: 209-398-2
UN: 1701
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR