4-Bromo-2-fluorotoluene ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Bromo-3-fluoro-4-methylbenzene

A15891.06 A15891.14 A15891.22
ALFAA15891.06EA 57.6 SGD
ALFAA15891.06 ALFAA15891.14 ALFAA15891.22
4-Bromo-2-fluorotoluene ≥99%
4-Bromo-2-fluorotoluene

Purity: 99% CAS No.: 51436-99-8 Molecular Formula: C7H6BrF Formula Weight: 189.03 Flash Point: 73°(163°F) Refractive Index: 1.5290 Boiling Point: 66-68°/8mm Beilstein Reference: 1859028 MDL No.: MFCD00013551

Formula: C₇H₆BrF
Boiling Pt: 66…68 °C (8 mmHg)
Density: 1,492
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013551
CAS Number: 51436-99-8
EINECS: 257-201-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR