Sodium benzoate ≥99%

Supplier: Thermo Fisher Scientific
Warning

A15946.36 A15946.0C A15946.0E A15946.30
ALFAA15946.36EA 0 SGD
ALFAA15946.36 ALFAA15946.0C ALFAA15946.0E ALFAA15946.30
Sodium benzoate ≥99%
Sodium benzoate

Purity: 99% CAS No.: 532-32-1 Molecular Formula: C7H5NaO2 Formula Weight: 144.11 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 300° Solubility: Soluble in water, alcohol Beilstein Reference: 3572467 Merck Reference: 14,8582 MDL No.: MFCD00012463

Formula: C₆H₅COONa
Melting Pt: >300 °C
Density: 1,44 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 14,8582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR