4-Chloro-3-nitrobenzyl alcohol 98

Supplier: Thermo Fisher Scientific

A16024.06 A16024.14
ALFAA16024.06EA 0 SGD
ALFAA16024.06 ALFAA16024.14
4-Chloro-3-nitrobenzyl alcohol 98
4-Chloro-3-nitrobenzyl alcohol

Purity: 98% CAS No.: 55912-20-4 Molecular Formula: C7H6ClNO3 Formula Weight: 187.58 Melting Point: 62-64° MDL No.: MFCD00007086

Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Melting Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00007086
CAS Number: 55912-20-4
EINECS: 259-901-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR