Bis(2-ethylhexyl)sebacate 95%

Supplier: Thermo Fisher Scientific

A16049.AK A16049.AU A16049.0J
ALFAA16049.AKEA 0 SGD
ALFAA16049.AK ALFAA16049.AU ALFAA16049.0J
Bis(2-ethylhexyl)sebacate 95%
Bis(2-ethylhexyl) sebacate

Purity: 95% CAS No.: 122-62-3 Molecular Formula: C26H50O4 Formula Weight: 426.68 Flash Point: 110°(230°F) Refractive Index: 1.4510 Boiling Point: 211-213°/1mm Merck Reference: 14,1251 MDL No.: MFCD00009497

Formula: C₂₆H₅₀O₄
Boiling Pt: 211…213 °C (1 mmHg)
Melting Pt: –55 °C
Density: 0,914
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009497
CAS Number: 122-62-3
EINECS: 204-558-8
Merck Index: 14,1251
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18b3fddd17e2c2f9148bc413e5e2c4558f3bc0a9&vid=689b9837fe6dd360b0805997dc4becc04db9113e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR