Zinc oxide heavy 98%

Supplier: Thermo Scientific
Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

A16188.0I
ALFAA16188.0IEA 0 SGD
ALFAA16188.0I
Zinc oxide heavy 98%
Zinc oxide

Purity: 98% CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Odour: Odourless Melting Point: 1975° UN No.: 3077 MDL No.: MFCD00011300

Formula: ZnO
MW: 81,39 g/mol
Melting Pt: 1975 °C
Density: 5,606
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 14,10147

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR