2-Bromo-4-chlorophenol ≥98%

Supplier: Thermo Fisher Scientific
Warning

A16218.14 A16218.22
ALFAA16218.14EA 0 SGD
ALFAA16218.14 ALFAA16218.22
2-Bromo-4-chlorophenol ≥98%
2-Bromo-4-chlorophenol

Purity: 98+% CAS No.: 695-96-5 Molecular Formula: C6H4BrClO Formula Weight: 207.46 Flash Point: 110°(230°F) Odour: Phenol-like Boiling Point: 230-232° Beilstein Reference: 2042871 MDL No.: MFCD00002319

Formula: C₆H₄BrClO
MW: 207.45 g/mol
Boiling Pt: 230…232 °C
Melting Pt: 31…35 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002319
CAS Number: 695-96-5
EINECS: 211-785-6
UN: 2020
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR