Sodium acetate trihydrate ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: Acetic acid sodium salt trihydrate

A16230.0C A16230.0E A16230.36
ALFAA16230.0CEA 678 SGD
ALFAA16230.0C ALFAA16230.0E ALFAA16230.36
Sodium acetate trihydrate ≥99%
Sodium acetate trihydrate

Purity: 99% CAS No.: 6131-90-4 Molecular Formula: NaOOCCH3.3H2O Formula Weight: 136.08 (82.03anhy) Odour: Odourless pH: 7.5-9.0 Melting Point: 62-64° Beilstein Reference: 3732037 Merck Reference: 14,8571 MDL No.: MFCD00071557

Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 62…64 °C
Density: 1.45
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR