4-Hydroxy-4-methyl-2-pentanone ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Diacetone alcohol

A16248.0F A16248.AE A16248.AP
ALFAA16248.0FEA 0 SGD
ALFAA16248.0F ALFAA16248.AE ALFAA16248.AP
4-Hydroxy-4-methyl-2-pentanone ≥98%
4-Hydroxy-4-methyl-2-pentanone

Purity: 98+% CAS No.: 123-42-2 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 58°(136°F) Refractive Index: 1.4230 Odour: Pleasant Melting Point: -47° Boiling Point: 168° Beilstein Reference: 1740440 Merck Reference: 14,2964 UN No.: 1148 MDL No.: MFCD00004471

Formula: (CH₃)₂C(OH)CH₂COCH₃
Boiling Pt: 168 °C
Melting Pt: –47 °C
Density: 0,935
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004471
CAS Number: 123-42-2
EINECS: 204-626-7
UN: 1148
ADR: 3,III
REACH: 01-2119473975-21
Merck Index: 14,2964
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2da78bff918d252787ee81ab8857bd3b19e51ac&vid=d8a0cccff2cac15b826ba733a013451ac11e3b51&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR