Cyclohexylbenzene ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Phenylcyclohexane

A16320.22 A16320.36
ALFAA16320.22EA 0 SGD
ALFAA16320.22 ALFAA16320.36
Cyclohexylbenzene ≥97%
Cyclohexylbenzene

Purity: 97+% CAS No.: 827-52-1 Molecular Formula: C12H16 Formula Weight: 160.26 Flash Point: 98°(208°F) Refractive Index: 1.5260 Odour: Mild Melting Point: 5-6° Boiling Point: 239-240° Beilstein Reference: 1906803 UN No.: 3082 MDL No.: MFCD00001451

Formula: C₁₂H₁₆
MW: 160.26 g/mol
Boiling Pt: 239…240 °C
Melting Pt: 5…6 °C
Density: 0.945
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001451
CAS Number: 827-52-1
EINECS: 212-572-0
UN: 3082
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR