4'-Bromo-3'-chloroacetanilide 98

Supplier: Thermo Fisher Scientific

A16547.14
ALFAA16547.14EA 0 SGD
ALFAA16547.14
4'-Bromo-3'-chloroacetanilide 98
4'-Bromo-3'-chloroacetanilide

Purity: 98% CAS No.: 22459-81-0 Molecular Formula: C8H7BrClNO Formula Weight: 248.51 Melting Point: 123-125° Beilstein Reference: 2967800

Formula: H₃CC(O)NH(C₆H₃BrCl)
MW: 248.51 g/mol
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00040848
CAS Number: 22459-81-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR