Benzothiazol-2-ol 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Hydroxybenzothiazole

A16648.06 A16648.14
ALFAA16648.06EA 0 SGD
ALFAA16648.06 ALFAA16648.14
Benzothiazol-2-ol 98
Benzothiazol-2-ol

Purity: 98% CAS No.: 934-34-9 Molecular Formula: C7H5NOS Formula Weight: 151.18 Melting Point: 136-140° MDL No.: MFCD00022868

Formula: C₇H₅NOS
MW: 151.18 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00022868
CAS Number: 934-34-9
EINECS: 213-281-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR