Benzothiazol-2-ol ≥98%

Supplier: Thermo Fisher Scientific
Warning

A16648.06 A16648.14
ALFAA16648.06EA 0 SGD
ALFAA16648.06 ALFAA16648.14
Benzothiazol-2-ol ≥98%
Benzothiazol-2-ol

Purity: 98% CAS No.: 934-34-9 Molecular Formula: C7H5NOS Formula Weight: 151.18 Melting Point: 136-140° MDL No.: MFCD00022868

Formula: C₇H₅NOS
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00022868
CAS Number: 934-34-9
EINECS: 213-281-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR