cis-1,2-Bis(diphenylphosphino)ethylene 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (Z)-1,2-Bis(diphenylphosphino)ethene

A16859.03 A16859.06
ALFAA16859.03EA 0 SGD
ALFAA16859.03 ALFAA16859.06
cis-1,2-Bis(diphenylphosphino)ethylene 97%
cis-1,2-Bis(diphenylphosphino)ethylene

Purity: 97% CAS No.: 983-80-2 Molecular Formula: C26H22P2 Formula Weight: 396.41 Melting Point: 122-126° Beilstein Reference: 757792 MDL No.: MFCD00003046

Formula: C₂₆H₂₂P₂
Melting Pt: 122…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00003046
CAS Number: 983-80-2
EINECS: 213-569-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR