(2-Bromophenyl)acetic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(2-Bromophenyl)acetic acid

A16900.06 A16900.14 A16900.22
ALFAA16900.06EA 0 SGD
ALFAA16900.06 ALFAA16900.14 ALFAA16900.22
(2-Bromophenyl)acetic acid ≥98%
(2-Bromophenyl)acetic acid

Purity: 98+% CAS No.: 18698-97-0 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 105-106° Beilstein Reference: 2250655

Formula: (C₆H₄Br)CH₂COOH
MW: 215.05 g/mol
Boiling Pt: 308
Melting Pt: 105…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00004314
CAS Number: 18698-97-0
EINECS: 242-509-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR